Orders LinkContactUsLinkComponent
Forceps
Straight, blunt, 250 mm
Catalog # 232-2117
Supplier:   RSG
Forceps
Catalog # 232-2117
Supplier:  RSG
Forceps
Forceps
Straight, blunt, 250 mm
Price
17,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
Specifications
  • Type:
    Straight, blunt
  • Length:
    250 mm