Tripods
With detachable legs, 120×210 mm
Catalog # 451-9106
Supplier: RSG
Jump to:
Product Details & Documents
Separable tripods made from stainless steel.
Specifications
- Int.Ø×H:120×210 mm
- Description:With detachable legs
- Seq:02
Accessories
Product Information
- Catalog #AvailabilityPrice
- Tripod extensions, set of three, total height up to 370 mmTripod extensions, set of three, total height up to 370 mmRSGA497.370Log in for availability1 * 3 ST - 68,50 €