Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

150 mm, 4 mm
Catalog # 231-1412
Supplier:   RSG
Double spatulas, micro
Catalog # 231-1412
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro

150 mm, 4 mm

Price
7,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Anti-magnetic, very flexible micro powder spatula.

  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance

Documents

Specifications
  • Length:
    150 mm
  • Blade width:
    4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.