Forceps with guide pin
Curved, with guide pin, 160 mm
Catalog # 232-2167
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
- With curved, extra fine tips
- Corrosion resistant
Specifications
- Type:Curved, with guide pin
- Length:160 mm