Order Entry
Belgium
Orders LinkContactUsLinkComponent
Support rings

Support rings

Heavy support ring, with clamp, open, with rubber coating, 100 mm
Catalog # RSGA284.100
Supplier:   RSG
Support rings
Catalog # RSGA284.100
Supplier:  RSG
Support rings
Support rings

Heavy support ring, with clamp, open, with rubber coating, 100 mm

Price
56,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

These heavy-duty support rings are made from stainless steel.

  • 8 mm rod
  • Available with rubber covered rings
Specifications
  • Ø int.:
    100 mm
  • Description:
    Heavy support ring, with clamp, open, with rubber coating
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.