Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Support rings

Heavy support ring, with rod, open, with rubber coating, 70 mm
Catalog # 241-0325
Supplier:   RSG
Support rings
Catalog # 241-0325
Supplier:  RSG
Support rings
Support rings

Heavy support ring, with rod, open, with rubber coating, 70 mm

Price
34,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

These heavy-duty support rings are made from stainless steel.

  • 8 mm rod
  • Available with rubber covered rings
Specifications
  • Ø int.:
    70 mm
  • Description:
    Heavy support ring, with rod, open, with rubber coating