Order Entry
Belgium
Orders LinkContactUsLinkComponent
Support rings

Support rings

Heavy support ring, with rod, open, with rubber coating, 50 mm
Catalog # 241-0324
Supplier:   RSG
Support rings
Catalog # 241-0324
Supplier:  RSG
Support rings
Support rings

Heavy support ring, with rod, open, with rubber coating, 50 mm

Price
33,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

These heavy-duty support rings are made from stainless steel.

  • 8 mm rod
  • Available with rubber covered rings
Specifications
  • Ø int.:
    50 mm
  • Description:
    Heavy support ring, with rod, open, with rubber coating
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.