Orders LinkContactUsLinkComponent
Scissors, multipurpose
Scissors, multipurpose
Laboratory, fine, 90 mm
Catalog # 233-1285
Supplier:   RSG
Scissors, multipurpose
Catalog # 233-1285
Supplier:  RSG
Scissors, multipurpose
Scissors, multipurpose
Laboratory, fine, 90 mm
Price
26,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, with black plastic handles.
  • Corrosion resistant
  • Available in fine or normal design, with large handles
  • Easy action
Specifications
  • Type:
    Laboratory, fine
  • Length:
    90 mm