Orders LinkContactUsLinkComponent
Microscopy instrument set
Microscopy instrument set
Microscopy set, 7 pcs.
Catalog # RSGA066.807
Supplier:   RSG
Microscopy instrument set
Catalog # RSGA066.807
Supplier:  RSG
Microscopy instrument set
Microscopy instrument set
Microscopy set, 7 pcs.
Price
158,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Microscopy instrument set in a grey wooden box, all articles are completely made of stainless steel.
  • 1 pair of microscopy scissors
  • 1 microscopy needle straight
  • 1 microscopy needle bent
  • 1 pair of tweezers, pointed straight with guide pin
  • 1 scalpel
Ordering information: On demand the arrangement of the 7 pcs. set can be individualised.
Specifications
  • Description:
    Microscopy set, 7 pcs.