Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Stand bases

200×315 mm, 3 kg, White
Catalog # 241-0183
Supplier:   R L ENTERPRISES
Stand bases
Catalog # 241-0183
Supplier:  R L ENTERPRISES
Stand bases
Stand bases

200×315 mm, 3 kg, White

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Rectangular retort bases with M10 threaded hole in centre of short side.

  • Steel, with acrylic finish
  • Acid resistant surface
  • Non-slip rubber feet for a firm footing

 : Supplied without rods which need to be ordered separately.

Specifications
  • Colour:
    White
  • L×W:
    200×315 mm
  • Seq:
    07
  • Weight:
    3 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.