Stand bases
200×315 mm, 3 kg, Blue
241-0182
Rectangular retort bases with M10 threaded hole in centre of short side.
- Steel, with acrylic finish
- Acid resistant surface
- Non-slip rubber feet for a firm footing
: Supplied without rods which need to be ordered separately.
- Colour:Blue
- L×W:200×315 mm
- Seq:08
- Weight:3 kg



