Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Stand bases

100×160 mm, 0,75 kg, Blue
Catalog # 241-0173
Supplier:   R L ENTERPRISES
Stand bases
Catalog # 241-0173
Supplier:  R L ENTERPRISES
Stand bases
Stand bases

100×160 mm, 0,75 kg, Blue

Price
50,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Rectangular retort bases with M10 threaded hole in centre of short side.

  • Steel, with acrylic finish
  • Acid resistant surface
  • Non-slip rubber feet for a firm footing

 : Supplied without rods which need to be ordered separately.

Specifications
  • Colour:
    Blue
  • L×W:
    100×160 mm
  • Seq:
    02
  • Weight:
    0,75 kg