Orders LinkContactUsLinkComponent
Small flanges with thread
Small flanges with thread
Small flanges KF, NW 16 with thread, G1/4", for ME 2, MZ 2, MV 2, DN 16
Catalog # 185-2277
Supplier:   VACUUBRAND
Small flanges with thread
Catalog # 185-2277
Supplier:  VACUUBRAND
Small flanges with thread
Small flanges with thread
Small flanges KF, NW 16 with thread, G1/4", for ME 2, MZ 2, MV 2, DN 16
Price
41,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These small flanges with thread made of stainless steel.
Documents
Specifications
  • Description:
    Small flanges KF, NW 16 with thread, G1/4", for ME 2, MZ 2, MV 2, DN 16
  • Seq:
    01