Face shields
Anti-UV (UV 380), anti-fog, Cellulose acetate, 195×540 mm
Catalog # 111-0222
Supplier: UVEX
Jump to:
Product Details & Documents
Lightweight face shields made from durable plastic with headrest and a 1 mm thick acetate screen.
- Integrated forehead protection
- Twist ratchet enables simple and reliable adjustment to fit most sizes of head
- Flip-up mechanism allows the visor to be set in any position
- Easy lens replacement
EN 166
Certifications: EN 166, EN 170
Specifications
- Type:Anti-UV (UV 380), anti-fog
- L×W:195×540 mm
- Material:Cellulose acetate
Accessories
Product Information
- Catalog #MaterialAvailabilityPrice
- Replacement visor lens
Product Information
- Catalog #MaterialAvailabilityPrice
- Replacement visor lens, anti-fogReplacement visor lens, anti-fog111-8229Cellulose acetateLog in for availability1 * 1 ST - 33,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



