Order Entry
Belgium
Orders LinkContactUsLinkComponent
Face shields

Face shields

Anti-UV (UV 380), Cellulose acetate, 195×412 mm
Catalog # 111-0221
Supplier:   UVEX
Face shields
Catalog # 111-0221
Supplier:  UVEX
Face shields
Face shields

Anti-UV (UV 380), Cellulose acetate, 195×412 mm

Price
31,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Lightweight face shields made from durable plastic with headrest and a 1 mm thick acetate screen.

  • Integrated forehead protection
  • Twist ratchet enables simple and reliable adjustment to fit most sizes of head
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement

EN 166

Certifications: EN 166, EN 170

Specifications
  • Type:
    Anti-UV (UV 380)
  • L×W:
    195×412 mm
  • Material:
    Cellulose acetate
Accessories

Product Information

  • Catalog #
    Material
    Availability
    Price
  • Replacement visor lens
    Replacement visor lens
    111-2543
    Cellulose acetate
    Log in for availability
    1 * 1 ST - 23,90 €

Product Information

  • Catalog #
    Material
    Availability
    Price
  • Replacement visor lens, anti-fog
    Replacement visor lens, anti-fog
    111-8229
    Cellulose acetate
    Log in for availability
    1 * 1 ST - 33,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.