Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dissecting needles

Dissecting needles

Curved, 140 mm
Catalog # USBE5480
Supplier:   USBECK Laborgeräte
Dissecting needles
Catalog # USBE5480
Dissecting needles
Dissecting needles

Curved, 140 mm

Price
1,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel needles with plastic handles.

Weight: 4 - 5 g

Specifications
  • Form:
    Curved
  • Length:
    140 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.