Orders LinkContactUsLinkComponent
Diamond pen
Diamond pen
Diamond pencil, ballpoint pen, 140 mm
Catalog # 201-0392
Supplier:   USBECK Laborgeräte
Diamond pen
Catalog # 201-0392
Diamond pen
Diamond pen
Diamond pencil, ballpoint pen, 140 mm
Price
40,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Diamond tip in metal holder.
  • Available with wooden handle or in the form of a ballpoint pen
  • Not suitable for engraving
  • 140 mm long
For labelling glass and metal surfaces.
Specifications
  • Description:
    Diamond pencil, ballpoint pen, 140 mm