Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Scoops

450 ml, 300 mm
Catalog # USBE3724
Supplier:   USBECK Laborgeräte
Scoops
Catalog # USBE3724
Scoops
Scoops

450 ml, 300 mm

Price
13,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Aluminium, smooth. For collecting or measuring powders, granulates or pastes.

  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    450 ml
  • Length:
    300 mm