Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Scoops

60 ml, 155 mm
Catalog # 231-0323
Supplier:   USBECK Laborgeräte
Scoops
Catalog # 231-0323
Scoops
Scoops

60 ml, 155 mm

Price
4,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Aluminium, smooth. For collecting or measuring powders, granulates or pastes.

  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    60 ml
  • Length:
    155 mm