Orders LinkContactUsLinkComponent
Blunt forceps
Blunt forceps
145 mm
Catalog # 232-0102
Supplier:   USBECK Laborgeräte
Blunt forceps
Catalog # 232-0102
Blunt forceps
Blunt forceps
145 mm
Price
4,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Nickel plated steel.
  • Curved
  • Blunt pointed ends
Documents
Specifications
  • Length:
    145 mm