Blunt forceps
115 mm
Catalog # 232-0100
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Nickel plated steel.
- Curved
- Blunt pointed ends
Documents
USBE3042_TDS_EN USBE3043_TDS_EN
Datasheet
Specifications
- Longueur:115 mm