Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
160 mm
Catalog # 232-0098
Supplier:   USBECK Laborgeräte
Forceps, straight
Catalog # 232-0098
Forceps, straight
Forceps, straight
160 mm
Price
4,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made from nickel plated steel or from stainless steel.
  • Straight
  • Fine points
Specifications
  • Length:
    160 mm