Orders LinkContactUsLinkComponent
Clamps, universal
Clamps, universal
Zinc die-cast, powder coated, 0 - 80 mm
Catalog # 241-7549
Supplier:   USBECK Laborgeräte
Clamps, universal
Catalog # 241-7549
Clamps, universal
Clamps, universal
Zinc die-cast, powder coated, 0 - 80 mm
Price
17,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
The fingers of the clamps have cork inserts to protect the clamped items.
  • Without bosshead for fixing to stand
  • With T-screws for rapid easy clamping
Specifications
  • Spanning width:
    0 - 80 mm
  • Material:
    Zinc die-cast, powder coated