Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stand plates

Stand plates

210×130 mm, 1,7 kg
Catalog # 241-7111
Supplier:   USBECK Laborgeräte
Stand plates
Catalog # 241-7111
Stand plates
Stand plates

210×130 mm, 1,7 kg

Price
24,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, powder coated. Firm, non slip stand. For a wide variety of uses. With M10 threaded hole on face side.

  • Rubber feet for a firm footing
  • Easy to clean
  • Robust
Specifications
  • L×W:
    210×130 mm
  • Weight:
    1,7 kg