Orders LinkContactUsLinkComponent
Burners
Burners
Teclu burner with needle valve, Natural gas
Catalog # 451-1365
Supplier:   USBECK Laborgeräte
Burners
Catalog # 451-1365
Burners
Burners
Teclu burner with needle valve, Natural gas
Price
64,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
DIN type.
  • Air regulation and pilot flame inside
Ordering information: Supplied without safety base, this needs to be ordered separately.
Specifications
  • Description:
    Teclu burner with needle valve
  • For:
    Natural gas