Orders LinkContactUsLinkComponent
Closures for headspace vials
Closures for headspace vials
Aluminium coated silicone cap/septa/spring, unassembled
Catalog # PERCB0104243
Supplier:   PerkinElmer
Closures for headspace vials
Catalog # PERCB0104243
Supplier:  PerkinElmer
Closures for headspace vials
Aluminium coated silicone cap/septa/spring, unassembled
Price
190,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
  • Description:
    Aluminium coated silicone cap/septa/spring, unassembled
  • Seq:
    04