Water testing kit, MINIKIT
Product Details & Documents
MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.
- MINIKITS are based on titrimetric methods
- Easy operation and exact reagent dosing
- Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years
Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.
Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:1 KIT
- Description:Hardness Total
- Type:AF 424
- Range:5 - 500 mg/l CaCO<sub>3</sub>, ~ 0,05 - 5 mmol/l
Specifications
Accessories
Product Information
- Catalog #AvailabilityPrice
- Dilution/shaker tube with lid, plastic, graduated, 100 mlDilution/shaker tube with lid, plastic, graduated, 100 mlLOVI385130Log in for availability1 * 1 ST - 9,90 €
Product Information
- Catalog #AvailabilityPrice
- Dilution/shaker tube with lid, plastic, graduated, 30 mlDilution/shaker tube with lid, plastic, graduated, 30 mlLOVI385132Log in for availability1 * 1 ST - 9,90 €
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.