Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water testing kit, MINIKIT

Hardness Total (Yes/No), Limit 4 mg/l, 8 mg/l or 20 mg/l CaCO3, ~ 0,04 or 0,08 or 0,2 mmol/l, AF 423, 1 KIT
Catalog # LOVI414230
Supplier:   Lovibond
Water testing kit, MINIKIT
Catalog # LOVI414230
Supplier:  Lovibond
CAS Number:  
Water testing kit, MINIKIT
Water testing kit, MINIKIT

Hardness Total (Yes/No), Limit 4 mg/l, 8 mg/l or 20 mg/l CaCO3, ~ 0,04 or 0,08 or 0,2 mmol/l, AF 423, 1 KIT

CAS Number:  
Price
63,90 €
Log in for availability
1 * 1 KIT
Product Details & Documents

MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.

  • MINIKITS are based on titrimetric methods
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years

Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.

Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.

Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.

 

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Hardness Total (Yes/No)
  • Type:
    AF 423
  • Range:
    Limit 4 mg/l, 8 mg/l or 20 mg/l CaCO<sub>3</sub>, ~ 0,04 or 0,08 or 0,2 mmol/l

Specifications

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 100 ml
    Dilution/shaker tube with lid, plastic, graduated, 100 ml
    LOVI385130
    Log in for availability
    1 * 1 ST - 9,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 30 ml
    Dilution/shaker tube with lid, plastic, graduated, 30 ml
    LOVI385132
    Log in for availability
    1 * 1 ST - 9,90 €