Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mesitylene for synthesis

Mesitylene for synthesis

Glass bottle, 1 L
Catalog # 8.05890.1000
Flammable
Health
Irritant
Environment
Supplier:   Merck
Mesitylene for synthesis
Catalog # 8.05890.1000
Supplier:  Merck
CAS Number:  108-67-8
Mesitylene for synthesis

Glass bottle, 1 L

CAS Number:  108-67-8
Price
87,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:165 °C (1013 hPa)
  • Melting Pt:–45 °C
  • Density:0.8637 g/cm³ (20 °C)
  • Flash Pt:48 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.