Tmb (Soluble Substrate) 1 * 1.000 mL
Catalog # CL07-1000
Supplier: Merck Millipore (Oncogene)
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF
Seals for ALPS™ 50 V heat sealer
Supplier: THERMO MATRIX TECHNOLOGIES
VWR®, Nitrile Gloves
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
96-Well Polypropylene Storage Blocks, Corning®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
VWR®, Deep Well Plates, Square Well
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Laemmli SDS sample buffer, reducing (6X)
Supplier: Thermo Fisher Scientific
Storage plate, 48-well, 6,0 ml
Supplier: THERMO MATRIX TECHNOLOGIES