Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Chlorophenol for synthesis

4-Chlorophenol for synthesis

Glass bottle, 1 kg
Catalog # 8.02254.1000
Irritant
Environment
Supplier:   Merck
4-Chlorophenol for synthesis
Catalog # 8.02254.1000
Supplier:  Merck
CAS Number:  106-48-9
4-Chlorophenol for synthesis

Glass bottle, 1 kg

CAS Number:  106-48-9
Price
94,60 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Merck Index:13,02173
  • Boiling Pt:220 °C (1013 hPa)
  • Melting Pt:41…44 °C
  • Density:1.31 g/cm³ (20 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • UN:2020
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-402-6

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Solubility:
    27 g/l (20 °C)
  • Melting Point:
    41 - 44 °C
  • Molar Mass:
    128.55 g/mol
  • Boiling Point:
    220 °C (1013 hPa)
  • Vapor Pressure:
    0.15 hPa (20 °C)
  • Flash Point:
    121 °C
  • Density:
    1.26 g/cm3 (45 °C)
  • Synonyms:
    4-Chloro-1-hydroxybenzene
  • Hill Formula:
    C6H5ClO
  • Chemical Formula:
    4-(Cl)C6H4OH

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.