4-Chlorophenol for synthesis
Glass bottle, 100 g
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: p-Chlorophenol
- CAS Number:106-48-9
- Formula:C₆H₅ClO
- MDL Number:MFCD00002318
- Molecular Weight:128.56 g/mol
- Merck Index:13,02173
- Boiling Pt:220 °C (1013 hPa)
- Melting Pt:41…44 °C
- Density:1.31 g/cm³ (20 °C)
- Flash Pt:121 °C
- Storage Temperature:Ambient
- UN:2020
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- EINECS:203-402-6
Documents
Specifications
Specification Test Results
- Solubility:27 g/l (20 °C)
- Melting Point:41 - 44 °C
- Molar Mass:128.55 g/mol
- Boiling Point:220 °C (1013 hPa)
- Vapor Pressure:0.15 hPa (20 °C)
- Flash Point:121 °C
- Density:1.26 g/cm3 (45 °C)
- Synonyms:4-Chloro-1-hydroxybenzene
- Hill Formula:C6H5ClO
- Chemical Formula:4-(Cl)C6H4OH
Specifications
- Pack type:Glass bottle
- Pk:100 g



