Order Entry
Belgium
Orders LinkContactUsLinkComponent
Containers with optional snap caps, Nunc™

Containers with optional snap caps, Nunc™

100 ml, 54 mm, Non sterile, without cap, Bag of 300, Carton of 300
Catalog # 391-8714
Containers with optional snap caps, Nunc™
Catalog # 391-8714
Supplier:  
Containers with optional snap caps, Nunc™
Containers with optional snap caps, Nunc™

100 ml, 54 mm, Non sterile, without cap, Bag of 300, Carton of 300

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Container sample 110ml with lid PP sterile (1/bag) 1 * 100 items Catalog #391-0020

Product 391-0020 is similar to 391-8714 but manufacturer and specifications may vary.

Product Details & Documents

PS, clear.

  • Transparent polystyrene construction
  • Versatile containers made of high-quality resins
  • Snap caps available for all containers
  • Cylindrical shape
  • Non sterile

Ordering information: Caps must be ordered seperately.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Packed:
    Bag of 300, Carton of 300
  • Type:
    Non sterile, without cap
  • Capacity:
    100 ml
  • Height:
    54 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.