Rosetta(De3)Plyss Comp Cells 1 * 0,4 mL
Catalog # 70956-3
Supplier: Merck Millipore (Novagen)
Frequently Bought Together
Carboys, narrow neck, PC, square, Biotainer™, Nalgene®
Supplier: Thermo Fisher Scientific
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: STERILIN
Internal thread cryogenic vials
Supplier: Corning
Vials for environmental analysis, WHEATON®
Supplier: DWK Life Sciences
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Sealing mats for assay and storage microplates, Axygen®
Supplier: Corning
384-well (HTS) microplates, non treated, BRANDplates®
Supplier: Brand
Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
Coliform water testing vials, wide mouth, Corning®
Supplier: Corning
Closures, PPCO, 13 mm
Supplier: Thermo Fisher Scientific
Sample containers with screw cap
Supplier: DELTALAB