Orders LinkContactUsLinkComponent
pH indicator strips, Whatman™
pH indicator strips, Whatman™
Universal indicator papers, non bleeding strips, 2,0 - 9,0
Catalog # WHAT10362010
pH indicator strips, Whatman™
Catalog # WHAT10362010
pH indicator strips, Whatman™
pH indicator strips, Whatman™
Universal indicator papers, non bleeding strips, 2,0 - 9,0
Price
22,60 €
Log in for availability
1 * 100 ST
Product Details & Documents
Universal indicator strips which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
Specifications
  • pH range:
    2,0 - 9,0
  • Description:
    Universal indicator papers, non bleeding strips