Orders LinkContactUsLinkComponent
Extraction thimbles
48×200 mm, 2,0 mm
  515-8949
Extraction thimbles
  515-8949
Extraction thimbles
Extraction thimbles
48×200 mm, 2,0 mm
289,00 €
1 * 25 ST
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
  • Int.Ø×H:
    48×200 mm
  • Seq:
    29
  • Thickness:
    2,0 mm