Order Entry
Belgium
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

30×80 mm, 1,5 mm
Catalog # 515-8923
Extraction thimbles
Catalog # 515-8923
Extraction thimbles
Extraction thimbles

30×80 mm, 1,5 mm

Price
111,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.

Specifications
  • Int.Ø×H:
    30×80 mm
  • Seq:
    12
  • Thickness:
    1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.