Orders LinkContactUsLinkComponent
Extraction unit and drying attachment, LiChrolut®
Extraction unit and drying attachment, LiChrolut®
Disposable fluoroplastic liners
Catalog # 1.19874.0001
Supplier:   Merck
Extraction unit and drying attachment, LiChrolut®
Catalog # 1.19874.0001
Supplier:  Merck
Extraction unit and drying attachment, LiChrolut®
Extraction unit and drying attachment, LiChrolut®
Disposable fluoroplastic liners
Price
235,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
All the individual steps associated with solid phase extraction can be carried out using the LiChrolut® extraction unit. This transparent and vacuum-suitable unit, made of glass, can be used to prepare up to 12 samples simultaneously.
  • Control of the vacuum via a manometer located at the front
  • Individual, easy setting of the various flow rates using valves
Delivery information: Supplied with 1 lid with 12 standard valves and seal, 1 glass chamber with gauge and vacuum valve, 12 standard stainless steel cannules, 1 collecting rack (base plate with 3 support rods, centre plate, top plate with 10 mm boring and 12 clamps), 1 rack for volumetric flasks, 1 rack for 16 mm test tubes and 1 rack for autosampler vials.
Specifications
  • Description:
    Disposable fluoroplastic liners
  • Seq:
    04