Orders LinkContactUsLinkComponent
Molecular models, Molymod®
Molecular models, Molymod®
Organic/anorganic chemistry molecular model
Catalog # 761-0110
Marketsource
Supplier:   SPIRING ENTERPRISES
Molecular models, Molymod®
Catalog # 761-0110
Marketsource
Molecular models, Molymod®
Molecular models, Molymod®
Organic/anorganic chemistry molecular model
Price
41,90 €
Log in for availability
1 * 1 SET
Product Details & Documents
Sets for different molecular models holding securely, even larger molecules.
Ordering information: Delivered with mounting instructions, list of contents, and examples of common molecules.
Specifications
  • Description:
    Organic/anorganic chemistry molecular model
  • Seq:
    007