Orders LinkContactUsLinkComponent
Test strips, phosphate, Reflectoquant®, Supelco®
Test strips, phosphate, Reflectoquant®, Supelco®
Phosphate test, 5 - 120 mg/l PO₄, Phosphormolybdenum blue, RQflex® 20, 50 Tests
Catalog # 1.16978.0001
Supplier:   Merck
Test strips, phosphate, Reflectoquant®, Supelco®
Catalog # 1.16978.0001
Supplier:  Merck
CAS Number:  
Test strips, phosphate, Reflectoquant®, Supelco®
Test strips, phosphate, Reflectoquant®, Supelco®
Phosphate test, 5 - 120 mg/l PO₄, Phosphormolybdenum blue, RQflex® 20, 50 Tests
CAS Number:  
Price
119,00 €
Log in for availability
1 * 50 Tests
Product Details & Documents
Test strips for the reflectometers RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Specifications
Specifications
  • Chemical method:
    Phosphormolybdenum blue
  • Pk:
    50 Tests
  • Description:
    Phosphate test
  • For unit:
    RQflex® 20
  • Range:
    5 - 120 mg/l PO₄
  • Cat. No.:
    1.16978.0001
  • M-Clarity quality level:
    MQ100
Specifications
Complementary Products
Product Information
  • Catalog #
    Description
    Model
    Availability
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log in for availability
    1 * 1 ST - 1 420,00 €