ContactUsLinkComponent
FRACTOGEL EMD COO- (M) WEAK CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returned after delivery 1 * 500 ml
Catalog # 1.16886.0500
FRACTOGEL EMD COO- (M) WEAK CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returned after delivery 1 * 500 ml
Catalog # 1.16886.0500