ContactUsLinkComponent
FRACTOGEL EMD SO3- (M) STRONG CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returne d after delivery 1 * 500 ml
Catalog # 1.16882.0500
FRACTOGEL EMD SO3- (M) STRONG CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returne d after delivery 1 * 500 ml
Catalog # 1.16882.0500