Crack sets for sample preparation, Spectroquant®, Supelco®
Catalog # 1.14687.0001
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Complete sets for digestion in a thermoreactor. The subsequent determination can be carried out in Spectroquant® test cells or reagent test sets. Crack Sets are easy to use and make sample preparation straightforward. For the preparation of dirty, coloured or samples with high levels of suspended material prior to using the appropriate Spectroquant® kit.
- Contains all reagents required for selected tests
- Easy to use - with step-by-step instructions
Documents
ProductPage_114687 ProductPage_114688 ProductPage_114963
Product specification
Specifications
Specifications
- Pk:100 Tests
- Description:Crack Set 10 Spectroquant®
- Cat. No.:1.14687.0001
- M-Clarity quality level:MQ100
- Parameters:Pb, Cd, Cr, Fe, Cu, Ni, Zn and P(total)
- Contents:Digestion reagent, acid, neutralising agent for pH adjustment.
- No. of digestions:100
- Model:Crack Set 10
- Method:Peroxodisulfate/acidic
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



