Orders LinkContactUsLinkComponent
Modular Storage Drawers
Modular Storage Drawers
Drawers only, 40,5 × 23 × 5,1 cm
Catalog # 720-0280
Supplier:   Simport Scientific
Modular Storage Drawers
Catalog # 720-0280
Supplier:  Simport Scientific
Modular Storage Drawers
Modular Storage Drawers
Drawers only, 40,5 × 23 × 5,1 cm
Price
173,00 €
Log in for availability
1 * 6 ST
Product Details & Documents
Storage drawers from PS. Permanent storage of up to 165 embedding rings or 250 cassettes per drawer.
  • Interlocking ridges for stacking
  • Identification labels included
Specifications
  • Description:
    Drawers only, 40,5 × 23 × 5,1 cm
  • Seq:
    01