Accessories for Spectroquant® NOVA
Transporting case, NOVA 30/60
: Merck
To document the analysis values, either a standard computer or a printer can be connected.
- The Multi-Agate II software can control data acquisition or simply record recorded analysis data
- The data records are archived here as raw data or imported into further programs such as Excel
Connectable photometers: SQ 118, SQ 200, SQ 300, Spectroquant® NOVA 30, Spectroquant® NOVA 60, Spectroquant® NOVA 400.
- Description:Transporting case
- Seq:901
- For:NOVA 30/60