Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, Certipur®, colour coded, Supelco®

Certipur® buffer solution, pH 7.00, green, Phosphate, Titripac®, 10 L
Catalog # 1.09477.9010
   Sustainable Packaging
Supplier:   Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Catalog # 1.09477.9010
Supplier:  Merck
   Sustainable Packaging
CAS Number:  
Buffer solutions, Certipur®, colour coded, Supelco®
Buffer solutions, Certipur®, colour coded, Supelco®

Certipur® buffer solution, pH 7.00, green, Phosphate, Titripac®, 10 L

CAS Number:  
Price
354,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C

Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).

Specifications

Specifications

  • Pack type:
    Titripac®
  • pH buffer type:
    Phosphate
  • Pk:
    10 L
  • Description:
    Certipur® buffer solution, pH 7.00, green
  • Cat. No.:
    1.09477.9010
  • M-Clarity quality level:
    MQ300
  • Environmentally Preferable:
    Sustainable Packaging

Specifications