Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, Certipur®, colour coded, Supelco®

Buffer solutions, Certipur®, colour coded, Supelco®

Certipur® buffer solution, pH 9.00, blue, Borate, Plastic bottle, 500 mL
Catalog # 1.09476.0500
Supplier:   Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Catalog # 1.09476.0500
Supplier:  Merck
CAS Number:  
Buffer solutions, Certipur®, colour coded, Supelco®
Buffer solutions, Certipur®, colour coded, Supelco®

Certipur® buffer solution, pH 9.00, blue, Borate, Plastic bottle, 500 mL

CAS Number:  
Price
59,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C

Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    500 mL
  • Description:
    Certipur® buffer solution, pH 9.00, blue
  • Cat. No.:
    1.09476.0500
  • M-Clarity quality level:
    MQ300

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.