Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, Certipur®, colour coded, Supelco®

Buffer solutions, Certipur®, colour coded, Supelco®

Certipur® buffer solution, pH 4,00, red, Citrate, Titripac®, 10 L
Catalog # 1.09475.9010
   Sustainable Packaging
Supplier:   Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Catalog # 1.09475.9010
Supplier:  Merck
   Sustainable Packaging
CAS Number:  
Buffer solutions, Certipur®, colour coded, Supelco®
Buffer solutions, Certipur®, colour coded, Supelco®

Certipur® buffer solution, pH 4,00, red, Citrate, Titripac®, 10 L

CAS Number:  
Price
354,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C

Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).

Specifications

Specifications

  • Pack type:
    Titripac®
  • pH buffer type:
    Citrate
  • Pk:
    10 L
  • Description:
    Certipur® buffer solution, pH 4,00, red
  • Cat. No.:
    1.09475.9010
  • M-Clarity quality level:
    MQ300
  • Environmentally Preferable:
    Sustainable Packaging

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.