Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sorbents, LiChrosorb®

Sorbents, LiChrosorb®

LiChrosorb® RP-8, 5 µm, 10 g
Catalog # 1.09332.0010
Supplier:   Merck
Sorbents, LiChrosorb®
Catalog # 1.09332.0010
Supplier:  Merck
CAS Number:  
Sorbents, LiChrosorb®
Sorbents, LiChrosorb®

LiChrosorb® RP-8, 5 µm, 10 g

CAS Number:  
Price
1 110,00 €
Log in for availability
1 * 10 g
Product Details & Documents

LiChrosorb® is an irregular shaped silica sorbent. LiChrosorb® packing materials offer the compete programme of non-polar derivatives (RP-8, RP-18, RP-select B) and polar derivatives (Si 60 and Si 100).

Specifications

Specifications

  • Particle Size:
    5 µm
  • Pk:
    10 g
  • Description:
    LiChrosorb® RP-8

Specifications