Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sorbents, LiChrosorb®

Sorbents, LiChrosorb®

LiChrosorb® Si 100, 10 µm, 10 g
Catalog # 1.09309.0010
Supplier:   Merck
Sorbents, LiChrosorb®
Catalog # 1.09309.0010
Supplier:  Merck
CAS Number:  
Sorbents, LiChrosorb®
Sorbents, LiChrosorb®

LiChrosorb® Si 100, 10 µm, 10 g

CAS Number:  
Price
235,00 €
Log in for availability
1 * 10 g
Product Details & Documents

LiChrosorb® is an irregular shaped silica sorbent. LiChrosorb® packing materials offer the compete programme of non-polar derivatives (RP-8, RP-18, RP-select B) and polar derivatives (Si 60 and Si 100).

Specifications

Specifications

  • Particle Size:
    10 µm
  • Pk:
    10 g
  • Description:
    LiChrosorb® Si 100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.